Learn More
Leica Biosystems Leica Snowcoat Microscope Slides, Pink - 1 x 3 in x 0.04 in, 1,440 slides/case
Supplier:  Leica Biosystems 3800340
Encompass
Pink Snowcoat Microscope Slides are pre-coated, color-coded glass slides designed for routine histology and pathology slide preparation. The Snowcoat marking area provides an easy surface for annotation and color-coded sample identification, and the slides accept xylene-resistant pens or pencils.
  • Pre-coated Snowcoat marking area for easy annotation.
  • Color-coded pink surface for quick sample identification.
  • Standard 1 x 3 in x 0.04 in slide dimensions.
  • Standard cut rectangular slides.
  • Sold 1,440 slides per case for bulk laboratory use.
  • Compatible with xylene-resistant pens and pencils.
Catalog No. NC3237843