Learn More
Medchemexpress LLC Pinobanksin (3,5,7-Trihydroxyflavanone) | 548-82-3 | 5 MG

Supplier: Medchemexpress LLC HYN30625MG
Pinobanksin, also known as 3,5,7-Trihydroxyflavanone, is a natural flavonoid derived from plants such as Pinus morrisonicola Hayata. This compound is recognized for its apoptotic induction properties in B-cell lymphoma cell lines.
- High purity of 99.16%.
- Molecular formula C15H12O5 and molecular weight 272.25.
- Appears as a white to off-white solid.
- Induces apoptosis in B-cell lymphoma cell lines.
- Inhibits melanogenesis and cell proliferation in B16-4A5 cells.
- Soluble in DMSO at 100 mg/mL.
- Multiple in vivo dissolution protocols available.
- Store at 4°C, protected from light.
- When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.