Learn More
Selleck Chemical LLC Pinocembrin S3941-5mg
SDP
Supplier:  Selleck Chemical LLC S39415MG
Encompass_Preferred
Pinocembrin (Dihydrochrysin Galangin flavanone 5 7-Dihydroxyflavanone) is a major flavonoid molecule incorporated as multifunctional in the pharmaceutical industry Its vast range of pharmacological activities has been well researched including antimicrobial anti-inflammatory antioxidant and anticancer activities
Catalog No. 50-310-8665