Learn More
TARGETMOL CHEMICALS INC Pinocembrin 7-O-3 -O-galloyl-4
SDP
Supplier:  TARGETMOL CHEMICALS INC T2S16825MG
Encompass_Preferred
Also available in 1 mg, 10 mg and bulk. Please contact Fisher for quotes. Pinocembrin 7-O-[3''-O-galloyl-4'',6''-hexahydroxydiphenoyl]-beta-D-glucoside can moderately inhibit alpha-amylase activity, with an IC50 value of 0.03 µmol/ml. Purity 98.73%
Catalog No. 50-287-9434