Learn More
Medchemexpress LLC Pinolenic acid | 16833-54-8 | 98.0% | 10 MG

Supplier: Medchemexpress LLC HY11954610MG
Pinolenic acid (PNLA) is a polyunsaturated fatty acid found in the seed oils of Pinus orientalis and Pinus pinaster. It is used in biochemical and pharmacological research for its reported anti-inflammatory and lipid-lowering activities and for studies of free fatty acid receptor signaling.
- Polyunsaturated fatty acid (CAS 16833-54-8).
- Purity 98.0%.
- Molecular formula C18H30O2; molecular weight 278.43 g/mol.
- Supplied in small research quantities (10 mg vial, 35.92 mM in ethanol for the 10 mg size).
- Documented with a product data sheet and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.