Learn More
Medchemexpress LLC Pinolenic acid | 16833-54-8 | 98.0% | 5 MG

Supplier: Medchemexpress LLC HY1195465MG
Pinolenic acid is a polyunsaturated fatty acid (PNLA) found in pine seed oils, reported to have anti-inflammatory and lipid-lowering activity. Supplied in small quantities as a solution for laboratory research; handle under appropriate ventilation and store frozen.
- Polyunsaturated fatty acid with reported anti-inflammatory activity.
- CAS number: 16833-54-8.
- Molecular formula: C18H30O2; molecular weight: 278.43 g/mol.
- Form: solution; recommended storage -20 °C.
- Intended for research use only; not for human or clinical use.
- Associated with free fatty acid receptor activity and GPCR/G protein pathway.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.