Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pinolenic acid ethyl ester | 493015-74-0 | MFCD12912314 | 98.0% | 306.48 g/mol | C20H34O2 | 10MG
SDP

Supplier:  Medchemexpress LLC HY12418710MG

Encompass_Preferred

Pinolenic acid ethyl ester is the ethyl ester derivative of pinolenic acid, a polyunsaturated fatty acid found in pine seed oils. It is supplied as a high-purity research reagent for biochemical and pharmacological studies, formulated as a liquid for use in lipid research, in vitro assays, and method development.

  • High purity suitable for research applications.
  • Liquid form, soluble in common organic solvents.
  • Available in small milligram packaging for experimental use.
  • Recommended storage: 4°C protected from light; in solvent -80°C up to 6 months, -20°C up to 1 month.
  • Molecular formula C20H34O2 and molecular weight 306.48 g/mol.
  • CAS number 493015-74-0 for unambiguous identification.

Catalog No. 50-002-36039


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.