Learn More
Cayman Chemical Pinolenic Acid methyl ester
SDP
Supplier:  Cayman Chemical 1000865610 MG
Encompass_Preferred
Pinolenic acid methyl ester is a neutral, more lipophilic form of the free acid.
Catalog No. 50-288-8992