Learn More
Cayman Chemical -PInoresInol 10mg
SDP
Supplier:  Cayman Chemical 2770010 MG
Encompass_Preferred
A lignan with diverse biological activities; an inhibitor of α-glucosidase and maltase (IC50s = 492 and 34.3 µM in Baker’s yeast and rat small intestine, respectively); scavenges ABTS, but not DPPH, radicals in cell-free assays (IC50s = 13.43 and >200 UG/ml, respectively); cytotoxic to A549, HepG2, and MCF-7, but not U251 and Bcap-37, cells (IC50s = 29.35, 62.35, 75.32, >80, and >80 µM, respectively); prevents cell death induced by glutamate in HT22 cells (EC50 = 6.96 µM) and inhibits LPS-induced nitric oxide production in RAW 264.7 cells (IC50 = 7.89 µM)
Catalog No. 50-293-7225