Learn More
Medchemexpress LLC Pinoresinol 4-O-β-D-glucopyranoside | 69251-96-3 | 99.9% | 520.53 g/mol | C26H32O11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN216810MG
Encompass_Preferred
Pinoresinol 4-O-β-D-glucopyranoside is a plant-derived lignan glycoside provided for research use. It is reported as an orally active α-glucosidase inhibitor and shows antioxidant and anti-inflammatory activities. Supplied as a solid for in vitro and in vivo studies; not for clinical use.
  • Plant-derived lignan glycoside.
  • Orally active α-glucosidase inhibitor (IC50 48.13 μg/mL).
  • High purity, 99.89%.
  • Supplied as a solid; store sealed, away from moisture and light.
Catalog No. 50-002-95516