Learn More
Medchemexpress LLC Pinosylvin monomethyl ether | 35302-70-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN30565MG
Encompass_Preferred
Pinosylvin monomethyl ether is a chemical compound primarily used for research due to its antibacterial and fungicidal properties. It appears as a white to off-white solid with a molecular weight of 226.27 and has demonstrated various cellular effects, including cytotoxicity and anti-inflammatory activity.
  • Exhibits antibacterial and fungicidal activity
  • Shows cytotoxicity against human A2780 cells (IC50 of 10 μg/mL)
  • Acts as an agonist at rat TRPA1 in HEK293 cells (EC50 of 3.5 μM)
  • Functions as an antagonist at human TRPV1 and rat TRPA1 in HEK293 cells
  • Reduces LPS-induced MCP1, IL6 expression, and nitric oxide production in mouse J774 cells
  • Inhibits PGE-2 production in LPS-stimulated RAW264.7 cells
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-20413