Learn More
Instech PinPort™ for fast access to externalized catheters. 22ga connector, white. 10 sterile pouches of 5.
SDP
Supplier:  Instech PNP3F2250
Encompass
The Instech PNP3F22-50 PinPort is a 22ga white access port designed for fast, aseptic connection to externalized catheters or tubing in rodent studies. Its 3mm port supports quick dosing or sampling using a matching PinPort injector while maintaining a closed, needle-free system to reduce infection risk and improve patency. Supplied as 10 sterile pouches of 5, it offers reliable manual access for short-term infusion and sampling workflows.
Features
  • 22ga white PinPort access port
  • Fast, aseptic catheter access
  • Closed, needle-free system
  • Use only with PNP3M injectors
  • Packaged 10 pouches of 5
  • EtO sterilized
Catalog No. NC0614285