Learn More
Cayman Chemical PIoglItazon-d4 1mg
SDP
Supplier:  Cayman Chemical 182591 MG
Encompass_Preferred
An internal standard for the quantification of pioglitazone by GC- or LC-MS
Catalog No. 50-296-6270