Learn More
Medchemexpress LLC Pioglitazone-d4 (alkyl) | 1134163-31-7 | 98.55% | 360.46 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY13956S11MG
Encompass_Preferred
Pioglitazone-d4 (alkyl) is a deuterium-labeled Pioglitazone, which is a potent and selective PPARγ agonist. It has high affinity binding to the PPARγ ligand-binding domain with EC50 of 0.93 and 0.99 μM for human and mouse PPARγ, respectively.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-50891