Learn More
Medchemexpress LLC Pioglitazone hydrochloride | 112529-15-4 | MFCD00865768 | ≥98.0% | C19H21N3O3S·HCl | 25 MG
Supplier: Medchemexpress LLC 25MGHY14601R
Pioglitazone hydrochloride is a potent and selective PPARγ agonist with EC50s of 0.93 and 0.99 μM for human and mouse PPARγ, respectively. It can be used in diabetes research.
- Potent and selective PPARγ agonist.
- EC50s of 0.93 and 0.99 μM for human and mouse PPARγ.
- Suitable for diabetes research.