Learn More
Grainger Pipe Marker Drain 5 in H 8 in W
Supplier:  Grainger 14H840
Encompass_Preferred
The Grainger 14H840 is an ASTM A36 carbon steel plate with 0.5 inch thickness, 48 inch width, and 96 inch length. This hot rolled steel plate has a tensile strength range of 58,000 - 79,800 psi and conforms to ASTM A36 specifications. The 14H840 carbon steel plate from Grainger can be used for a variety of structural applications that require durability, strength, and weldability. With weight of 494 pounds per plate, this A36 steel meets chemistry and mechanical requirements and has good formability and strength.
Catalog No. 50-275-3249