Learn More
Grainger Pipe Marker Drain 9 in H 8 in W
Supplier:  Grainger 14H837
Encompass_Preferred
The Brady Pipe Marker Drain is a self-adhesive vinyl pipe marker that identifies drain pipes. It has a legend that reads "Drain" and comes in 1-1/2" height x 9" length rolls. The pipe markers are flexible, conforming to irregular surfaces and staying secured even in harsh conditions. They feature a strong acrylic adhesive that adheres to PVC, ABS, fiberglass and other low surface energy plastics. The vinyl material and adhesive resist UV light, chemicals, abrasion and moisture. This versatile pipe marker improves safety and meets ANSI/OSHA requirements for pipe labeling.
Catalog No. 50-274-9026