Learn More
Grainger Pipe Marker Natural Gas 9 in H 8 in W
Supplier:  Grainger 14H924
Encompass_Preferred
The Brady Natural Gas Pipe Marker 14H924 is a self-adhesive vinyl label designed for identifying natural gas pipes. It features the words "Natural Gas" printed in black on a yellow background. The marker has an aggressive adhesive that adheres to PVC, ABS, fiberglass, and painted metal pipes. It measures 4" high x 12" long and comes in a pack of 10 markers. The markers are weather, UV, chemical, and abrasion resistant. They can withstand temperatures from -40°F to 176°F. The markers comply with ANSI Z535.1 safety standards for facility safety signs and labels.
Catalog No. 50-275-1184