Learn More
Grainger Pipe Marker Vent 26 in H 12 in W
Supplier:  Grainger 14J009
Encompass_Preferred
The Brady Pipe Marker Vent 14J009 is a self-adhesive vinyl pipe marker designed for identifying ventilation pipes. It has a legend that reads "Vent" in black text on a yellow background. The marker measures 1-1/2" H x 9" L and is made from 6 mil vinyl with a permanent acrylic adhesive backing. It can withstand temperatures from -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides a neat, professional method of labeling ventilation pipes in commercial and industrial settings.
Catalog No. 50-275-1077