Learn More
Selleck Chemical LLC Pipecolic acid
SDP
Supplier:  Selleck Chemical LLC S63221G
Encompass_Preferred
Pipecolic acid (piperidine-2-carboxylic acid) a metabolite of lysine found in human physiological fluids such as urine plasma and CSF is an important regulator of immunity in plants and humans alike
Catalog No. 50-000-03687