Learn More
Selleck Chemical LLC Pipemidic acid S5051-25mg
SDP
Supplier:  Selleck Chemical LLC S505125MG
Encompass_Preferred
Pipemidic acid (Acido pipemidico) is a pyridopyrimidine antibiotic derivative of piromidic acid with activity against gram-negative bacteria including Pseudomonas aeruginosa and some gram-positive bacteria Pipemidic acid is an inhibitor of DNA gyrase that is a bacterial enzyme which catalyzes the ATP-dependent negative supercoiling of DNA It can be used for the research of intestinal urinary and biliary tract infections
Catalog No. 50-311-1895