Learn More
Medchemexpress LLC Pipendoxifene hydrochloride | 245124-69-0 | 99.8% | 493.04 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13724B100MG
Encompass_Preferred
Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) developed for breast cancer research. It exhibits an excellent preclinical pharmacological profile and is being developed as a treatment for metastatic breast cancer.
  • Selective estrogen receptor modulator (SERM)
  • Used for breast cancer research
  • Exhibits excellent preclinical pharmacological profile
  • Target for estrogen receptor
Catalog No. 50-003-50177