Learn More
Medchemexpress LLC Pipequaline | 77472-98-1 | MFCD00866983 | 99.9% | 316.44 g/mol | C22H24N2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100140100MG
Encompass_Preferred
Pipequaline (PK-8165) is a research chemical characterized as a partial benzodiazepine receptor agonist with anxiolytic activity. It is intended for preclinical investigations of GABAA/benzodiazepine receptor modulation and related neuropharmacology. Supplied as a high-purity solid for laboratory use; not for human or veterinary administration.
  • Partial benzodiazepine receptor agonist with anxiolytic activity.
  • Suitable for preclinical pharmacology and receptor binding studies.
  • High purity for reproducible experimental results.
  • Available in small milligram pack sizes to support dose-ranging work.
  • Molecular weight 316.44 g/mol, enabling stoichiometric calculations.
Catalog No. 50-002-15836