Learn More
Medchemexpress LLC Piperacillin | 61477-96-1 | 98.1% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB192310MM/1ML
Encompass_Preferred
Piperacillin is a semisynthetic broad-spectrum β-lactam antibiotic. It exhibits potent bactericidal activity against Gram-negative bacteria and selected Gram-positive strains by targeting penicillin-binding proteins. It is commonly combined with the β-lactamase inhibitor Tazobactam for enhanced efficacy.
  • Semisynthetic broad-spectrum β-lactam antibiotic
  • Potent bactericidal activity against Gram-negative bacteria
  • Effective against selected Gram-positive strains
  • Functions by targeting penicillin-binding proteins
  • Frequently used in combination with β-lactamase inhibitors
  • Demonstrates in vitro inhibition against various bacteria including Pseudomonas aeruginosa and Escherichia coli
  • Prolongs survival in animal models of infection when combined with Tazobactam
Catalog No. 50-004-11779