Learn More
Medchemexpress LLC Piperacillin sodium | 59703-84-3 | 98.22% | 200 MG
SDP
Supplier:  Medchemexpress LLC HYB1286200MG
Encompass_Preferred
Piperacillin sodium is a semisynthetic broad-spectrum β-lactam antibiotic that exhibits potent bactericidal activity against Gram-negative bacteria as well as select Gram-positive strains through penicillin-binding proteins. It is most commonly used in combination with the β-lactamase inhibitor Tazobactam.
  • Exhibits potent bactericidal activity
  • Effective against Gram-negative bacteria
  • Effective against select Gram-positive strains
  • Targets penicillin-binding proteins
  • Commonly used with β-lactamase inhibitor
Catalog No. 50-004-13612