Learn More
Medchemexpress LLC 2-Piperazinone | 5625-67-2 | 99.97% | 100.12 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY20128
Encompass_Preferred
Piperazin-2-one is a piperazine ring derivative containing amide and ester carbonyl groups. It serves as an important intermediate that can be used for the synthesis of various active compounds. Piperazin-2-one derivatives have also demonstrated anti-adenoviral activity.
  • Is a piperazine ring derivative containing amide and ester carbonyl groups.
  • Can be used for the synthesis of various active compounds.
  • Derivatives also exhibit anti-adenoviral activity.
Catalog No. 50-004-02975