Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperazin-2-one-3,3,5,5,6,6-d6 | 1219803-71-0 | 99.3% | 106.16 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY20128S1MG

Encompass_Preferred

Piperazin-2-one-d6 is the deuterium labeled version of Piperazin-2-one, which is a piperazine ring derivative containing amide and ester carbonyl groups. It serves as an important intermediate for the synthesis of various active compounds. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.

  • Deuterium labeled compound
  • Stable heavy isotope
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Purity: 99.3%
  • Appearance: Solid, white to off-white

Catalog No. 50-004-04893


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.