Learn More
Medchemexpress LLC Piperazin-2-one-3,3,5,5,6,6-d6 | 1219803-71-0 | 99.3% | 106.16 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY20128S
Encompass_Preferred
Piperazin-2-one-d6 is the deuterium labeled Piperazin-2-one. Piperazin-2-one is a piperazine ring derivative containing amide and ester carbonyl groups. As an important intermediate, it can be used for the synthesis of various active compounds. It is for research use only and not sold to patients.
  • Stable heavy isotope of hydrogen, carbon, and other elements incorporated into drug molecules.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-02274