Learn More
Medchemexpress LLC Piperazine, 2-(chloromethyl)-1,4-bis(phenylmethyl)-, (2r)- | 2225787-91-5 | 99.1% | 314.86 | 100 MG

Supplier: Medchemexpress LLC HY23565100MG
(R)-1,4-Dibenzyl-2-(chloromethyl)piperazine is a drug intermediate used for the synthesis of various active compounds. This product is for research use only and not sold to patients.
- Molecular weight: 314.86
- Formula: C19H23ClN2
- CAS number: 2225787-91-5
- Appearance: Oil
- Color: Colorless to light yellow
- Purity: 99.10%
- Shipping at room temperature in continental US
- Storage at 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture).
- Suitable for synthesis of various active compounds.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.