Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 | C35H41ClN6O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1008875MG
Encompass_Preferred
Piperazine erastin is a synthetic analog of erastin that induces ferroptosis, an iron-dependent form of non-apoptotic cell death. It is supplied as a purified solid for research use in cellular and in vivo studies probing ferroptosis mechanisms, oxidative stress, and cancer-related cell death pathways.
  • Induces ferroptosis in cell-based assays.
  • High purity suitable for biochemical and cellular experiments.
  • Solid powder with defined storage and stability profiles.
  • Readily soluble in DMSO for formulation and dosing.
  • Available in small quantities for research-scale studies.
Catalog No. 50-002-11849