Learn More
Medchemexpress LLC Piperazine erastin | 1538593-71-3 | 99.1% | 645.19 g/mol | C35H41ClN6O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10088710MM 1ML
Encompass_Preferred
Piperazine erastin is a research reagent that induces ferroptosis, an iron-dependent form of non-apoptotic cell death. It is used in cellular and cancer research to investigate ferroptotic mechanisms; the piperazine substitution improves water solubility and metabolic stability relative to erastin.
  • Induces ferroptosis in cell models.
  • Improved water solubility and metabolic stability compared with erastin.
  • Available as a ready-to-use 10 mM solution in DMSO (1 mL).
  • Also available as a solid in multiple milligram quantities for flexibility.
  • High purity, typically 99.12% as reported by the manufacturer.
  • Useful for cancer research and studies of iron-dependent cell death pathways.
Catalog No. 50-002-12920