Learn More
TARGETMOL CHEMICALS INC PIPERAZINE ERASTIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T165405MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Piperazine erastin is an analog of erastin. It causes an iron-dependent form of non-apoptotic cell death termed ferroptosis. purity: 97%
Catalog No. 50-258-1167