Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Piperic acid | 136-72-1 | 97.0% | 100 MG
SDP

Supplier:  Medchemexpress LLC HY121909100MG

Encompass_Preferred

Piperic acid is a research-grade antioxidant and antibacterial compound (CAS 136-72-1) used in studies of food preservation and human health. It is supplied as a light yellow solid at high purity for laboratory research.

  • Purity 97.0%.
  • Molecular weight 218.21 g/mol.
  • Chemical formula C12H10O4.
  • Available as a 100 MG solid for laboratory use.
  • Storage: powder stable at -20°C for up to 3 years.

Catalog No. 50-002-31085


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.