Learn More
Medchemexpress LLC Piperidin-4-d5 | 1219799-35-5 | 106.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW004328S5MG
Encompass_Preferred
Piperidin-4-ol-d5 is a deuterium-labeled derivative of Piperidin-4-ol, intended for research purposes only and not for human use. This compound can serve as a tracer and an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes into drug molecules, is primarily used for quantitation during drug development and has shown potential to influence the pharmacokinetic and metabolic profiles of drugs. The product appears as a white to off-white solid.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-46273