Learn More
Medchemexpress LLC Piperidine-4-sulfonic acid | 72450-62-5 | 99.9% | 165.21 | 25 MG

Supplier: Medchemexpress LLC 25MGHY139116
Piperidine-4-sulfonic acid is a potent GABA agonist with an IC50 of 0.034 μM for the inhibition of H-GABA binding. It is intended for research use only.
- Potent GABA agonist
- IC50 of 0.034 μM for inhibition of H-GABA binding
- Solid appearance, light brown to brown color
- Purity of 99.95%
- Stable in powder form at -20°C for 3 years or 4°C for 2 years
- Stable in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.