Learn More
Medchemexpress LLC Piperitone | 89-81-6 | 99.89% | 152.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN9496100MG
Encompass_Preferred
Piperitone is described as a powerful repellent and antiappetent agent. It is highly toxic to Cymbopogon schoenanthus adults, newly laid eggs, and neonate larvae, demonstrating insecticidal activity. It has also shown toxicity to Callosobruchus maculatus adults with an LC50 value of 1.6 μL/L and inhibits the development of newly laid eggs and neonate larvae, with strong ovicidal activity.
  • Acts as a powerful repellent and antiappetent agent.
  • Shows insecticidal activity against Cymbopogon schoenanthus.
  • Toxic to Callosobruchus maculatus adults.
  • Inhibits development of newly laid eggs and neonate larvae.
  • Exhibits strong ovicidal activity.
Catalog No. 50-004-26123