Learn More
Medchemexpress LLC Piperitone | 89-81-6 | MFCD00045532 | 99.89% | 152.24 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN9496500MG
Encompass_Preferred
Piperitone is a powerful repellent and antiappetent agent that exhibits insecticidal activity. It is highly toxic to *Cymbopogon schoenanthus* adults, newly laid eggs, and neonate larvae. It has been shown to be more toxic than crude oil to *Callosobruchus maculatus* adults, inhibiting the development of newly laid eggs and neonate larvae.
  • Powerful repellent and antiappetent agent
  • Exhibits insecticidal activity
  • Highly toxic to adults, newly laid eggs, and neonate larvae
  • More toxic than crude oil to insect adults
  • Inhibits development of newly laid eggs and neonate larvae
  • Strong ovicidal activity, aborting all eggs at 6.7 μL/L
  • Kills all neonate larvae at 10 μM
Catalog No. 50-004-26336