Learn More
Medchemexpress LLC Piperonyl alcohol | 495-76-1 | MFCD00005836 | 99.8% | 152.15 g·mol-1 | C8H8O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02810810G
Encompass_Preferred
Piperonyl alcohol (1,3-benzodioxole-5-methanol) is a benzodioxole-derived benzyl alcohol supplied for laboratory research. It is described as biocompatible and can be used as an initiator or building block in chemical synthesis. The product is offered as a solid and as pre-prepared DMSO solutions for assay convenience.
  • High reported purity (99.78% LCMS).
  • Solid, white to off-white physical form.
  • Soluble in DMSO (100 mg/mL; ultrasonic assistance recommended).
  • In vivo formulation protocols provided with solubility ≈2.5 mg/mL.
  • Available in multiple pack sizes including 10 g.
  • Intended for research use only.
Catalog No. 50-003-00139