Learn More
Medchemexpress LLC Piperonyl butoxide | 51-03-6 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB119810MM/1ML
Encompass_Preferred
Piperonyl butoxide is a pesticide synergist and food additive. It has been observed to have adverse effects on reproduction, development, and behavior in mice. Additionally, it can activate c-Jun and ATF-2 in mouse hepatocytes and is identified as a liver cancer carcinogen in rats and mice.
  • Induces phosphorylation of ATF-2 in mouse hepatocytes.
  • Elevates liver nuclear proteins that bind to an AP-1 consensus oligonucleotide.
  • Induces c-Jun phosphorylation.
  • Reduces average speed in exploratory behavior examination.
  • Shortens total exploration distance and decreases average speed in exploratory behavior examination.
Catalog No. 50-004-12232