Learn More
Medchemexpress LLC Piperonylic acid | 94-53-1 | 99.9% | 166.13 | 100 G
SDP
Supplier:  Medchemexpress LLC HY41404100G
Encompass_Preferred
Piperonylic acid is a natural molecule that structurally mimics trans-cinnamic acid due to its methylenedioxy function. It acts as a selective, mechanism-based inactivator of trans-cinnamate 4-Hydroxylase. This compound possesses anticancer, antioxidant, and antibacterial properties.
  • Mimics the structure of trans-cinnamic acid.
  • Functions as a selective, mechanism-based inactivator of trans-cinnamate 4-Hydroxylase.
  • Exhibits anticancer, antioxidant, and antibacterial activities.
  • Promotes growth and survival of HaCaT cells.
  • Accelerates wound healing in mice.
Catalog No. 50-004-04917