Learn More
Alkali Scientific PIPES Sesquisodium Salt [Piperazine N,N' bis (2 ethanesulfonic acid) sesquisodium salt], 500 G
Supplier: Alkali Scientific PES004CP
PIPES Sesquisodium Salt (Piperazine N,N' bis(2 ethanesulfonic acid) sesquisodium salt) in a 500G pack is a highly stable and versatile reagent used for preparing buffers in biochemical and laboratory applications. PIPES is an effective buffering agent that ensures a constant pH range ideal for a variety of biological and chemical processes. Its unique structure offers excellent pH stability, particularly in acidic conditions, making it suitable for sensitive biological experiments. PIPES is widely used in molecular biology, protein purification, and enzyme activity studies. It is especially beneficial for applications that require precise pH control over prolonged periods, such as electrophoresis and chromatography.