Learn More
Research Products International Corp Extra Long Tip, 1000ul, Non-Sterile, Bulk, 1000 per Case
SDP
Supplier:  Research Products International Corp 141216
Encompass
Our filter pipet tips effectively trap aerosol contaminates when using air displacement pipettors. Ideal for a wide variety of applications including DNA amplification, forensics, tissue culture and protocols that involve pipetting radioactive solutions. The special inert filter is non-self sealing and allows sample recovery in event of over pipetting. Tips are designed for Biohit Proline, Eppendorf,100-1000ul, P1000, Socorex 811, 821, 822.
Catalog No. 50-212-225