Learn More
Integra Biosciences Corp PIPETBOY acu 2, Pink Sunrise
SDP
Supplier:  Integra Biosciences Corp 155024
Encompass
The second generation of PIPETBOY acu is equipped with the latest technology providing an increased pipetting speed and a significant weight reduction of the instrument. These two features help you to cut your work time and offer you an improved ergonomic pipetting experience. Besides these changes the main benefits of PIPETBOY acu were preserved: the unmatched pipetting precision, the colorful look and the weight balance.
Catalog No. NC1834009