Learn More
Pipette Supplies, Inc. PIPETMAN FILTERS 5/10ML 100PK
SDP
Supplier:  Pipette Supplies, Inc. GI F161241
Encompass
Bag of 100 polyethylene filters for:Gilson Pipetman Classic 5000μL, 10mL (P5000, P10mL)Gilson Pipetman G 5000μL, 10mL (P5000G, P10mLG)Gilson Pipetman L 5000μL, 10mL (P5000L, P10mLL)Gilson Pipetman Fixed F5000Gilson Pipetman Ultra 5000μL, 10mL (U5000, U10mL)Gilson Pipetman Concept 5000μL, 10mL (C5000, C10mL)Rainin Classic 5000μL, 10mL (PR5000, PR10mL)Alternative to Rainin 17001944, 17001945.Made in France
Catalog No. 50-131-7636