Learn More
Pipette Supplies, Inc. PIPETMAN G/L LUBRICANT
SDP
Supplier:  Pipette Supplies, Inc. GI 5440011070/DEL
Encompass
Lubricant tube for the Gilson Pipetman G and Pipetman L single and multichannel pipettes.Now also available in a larger 100g jar size, GI FA07093.Made in France
Catalog No. 50-131-7638