Learn More
MARSHALL SCIENTIFIC Gilson PIPETMAN G Micro Kit
SDP
Supplier:  MARSHALL SCIENTIFIC F167800
Encompass_Preferred
The PIPETMAN G Micro Kit provides the three pipettes most used by molecular biologists for pipetting enzymes, reagents, and buffers - a P2G, P10G, and P100G - plus pipette holders, tips, and pipetting guide. With a redesigned piston assembly, the PIPETMAN G pipettes feature extremely low pipetting forces, improving comfort and minimizing injury. This is all accomplished while maintaining the durability, accuracy, and reliability of the original PIPETMAN pipettes.
Catalog No. NC2888825