Learn More
Pipette Supplies, Inc. PIPETMAN SHAFT P1000 1000UL
SDP
Supplier:  Pipette Supplies, Inc. 61000
Encompass
Tip Holder / shaft for:Gilson Pipetman Classic 1000μL (P1000)Rainin Classic 1000μL (PR1000)Gilson Pipetman F Fixed F250, F300, F400, F500, F1000Gilson Pipetman Neo 1000μL (P1000N)Gilson Pipetman G 1000μL (P1000G)Gilson Pipetman L 1000μL (P1000L)Alternative to Gilson F123371, F144498, F161835 & Rainin 23371, 17004984.Autoclavable.Made in the USA
Catalog No. 50-131-7603