Learn More
CELLTREAT Scientific Products 1-10uL Single Channel Pipette, PVDF, PTFE, Stainless Steel
SDP
Supplier:  CELLTREAT Scientific Products 230299
Encompass_Preferred
High-precision adjustable pipette with easy-to-read volume display. Continuously adjustable with a locking ring to prevent volume changes while pipetting. Lightweight design and easy operation with either handReduced operating forces; plunger is easy to press and dispense. Stainless steel, PTFE, and PVDF components for long life. It is recommended not to autoclave more than 6 times. Fits CELLTREAT Scientific Products Universal Pipette Tips for accurate pipetting.
Catalog No. NC1964612