Learn More
Chemglass Life Sciences Chemglass Pipette Tip, 1000 μL Sterile Barrier, Bulk Packed (CLS-3499D-1XL)
SDP
Supplier:  Chemglass Life Sciences CLS3499D1XL
Encompass_Preferred
Sterile 1,000 μL barrier pipette tips designed for precise molecular biology and analytical liquid handling. The ultra-clear, fine-point design minimizes sample retention and delivers accurate aspiration and dispensing. Hydrophobic barrier prevents aerosol carry-over and protects pipettors; tips are gamma-irradiated and certified free of RNase, DNase, human DNA, and pyrogens. Packaged bulk for high-throughput workflows.
  • Sterile, gamma-irradiated
  • 1000 μL volume
  • 100% polypropylene
  • Certified RNase-free, DNase-free, human DNA-free, and pyrogen-free
  • Hydrophobic aerosol barrier to minimize carry-over
  • Graduated for approximate volume checking
  • Universal fit for most single and multichannel pipettors
  • Packaging: 4,800 per case
Catalog No. 50-273-3215