Learn More
Chemglass Life Sciences Chemglass pipette tips, sterile, 20 μL standard barrier - fine-point, universal fit (4,800/case)
SDP
Supplier:  Chemglass Life Sciences CLS3499D020
Encompass_Preferred
Sterile 20 μL standard barrier pipette tips provide accurate low-volume dispensing while protecting pipettors from aerosol contamination. The low-retention, fine-point design and ultra-clear polypropylene construction make these tips suitable for sensitive nucleic acid and protein applications.
  • Tip volume: 20 μL.
  • Tip type: Standard barrier tip.
  • Material: Premium-grade 100% polypropylene.
  • Sterility: Sterile and gamma-irradiated.
  • Certified free of RNase, DNase, human DNA, and pyrogens.
  • Pack size: 4,800 per case.
  • Design features: Low-retention fine-point, graduated markings, ultra-clear polymer, and universal fit for most single and multichannel pipettors.
  • Aerosol protection: Hydrophobic barrier reduces carry-over and pipettor contamination.
Catalog No. 50-273-3217